Order Entry
Slovakia
ContactUsLinkComponent
 
Organic reference standard, D-(-)-Salicin, analytical standard, Supelco®
  79588-100MG
 :  Merck
 :  
undefined
Organic reference standard, D-(-)-Salicin, analytical standard, Supelco®
  79588-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    138-52-3
  • Description:
    D-(-)-Salicin, ≥98,5% (HPLC)
  • Formula:
    C₁₃H₁₈O₇
  • Description:
    D-(-)-Salicin
  • Form:
    Neat