Order Entry
Slovakia
ContactUsLinkComponent
 
Phytochemical standard, Cynarin, analytical standard, Supelco®
  91801-5MG
 :  Merck
 :  
undefined
Phytochemical standard, Cynarin, analytical standard, Supelco®
  91801-5MG
 :  Merck
 :  

 

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    30964-13-7
  • Description:
    Cynarin, ≥98,0% (HPLC)
  • Formula:
    C₂₅H₂₄O₁₂
  • Storage temperature:
    2...8 °C
  • Description:
    Cynarin
  • Form:
    Neat

 

 

Phenolic compound found in artichoke. Antioxidant and choleretic properties. Potential immunosuppressive agent.

 : Bottomless glass bottle. Contents are inside inserted fused cone.