Supelco® Organic Reference Standard, Cyanidin chloride
94099-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:528-58-5
- Description:Cyanidin chloride
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD00017582
- M-Clarity quality level:MQ100
- Synonym(s):3,3′,4,5,7-Pentahydroxyflavylium chloride; Cyanidol chloride
- Molecular weight:322,70