Phytochemical standard, cyanidin 3-sophoroside chloride, analytical standard, Supelco®
Catalog # 42739-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:18376-31-3
- Description:Cyanidin 3-sophoroside chloride, ≥85,0% (HPLC)
- Formula:C₂₇H₃₁ClO₁₆
- Storage temperature:–20 °C
- Description:Cyanidin 3-sophoroside chloride