Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Corosolic acid
Supelco® Organic Reference Standard, Corosolic acid
  89067-10MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Corosolic acid
  89067-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    4547-24-4
  • Description:
    Corosolic acid
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD06794973
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (2α,3β)-2,3-Dihydroxyurs-12-en-28-oic acid; 2α-Hydroxyursolic acid; Colosolic acid; Corsolic acid; Glucosol
  • Molecular weight:
    472,70