Phytochemical standard, cirsilineol, analytical standard, Supelco®
Catalog # 67403-5MG
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:41365-32-6
- Description:Cirsilineol, ≥95,0% (HPLC and enzymatic)
- Formula:C₁₈H₁₆O₇
- Description:Cirsilineol
- Form:Neat
Specifications
About this item
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.