Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Catechin gallate solution
Supelco® Organic Reference Standard, Catechin gallate solution
Catalog # 49061-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Catechin gallate solution
Catalog # 49061-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml in methanol, ampule of 0,5 ml
  • Pk:
    1
  • Standards:
    Analytical standard
  • Description:
    Catechin gallate solution
  • Storage conditions:
    –10...–25 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H370
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100

Specifications

About this item

  • GHS signal: GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311