Supelco® Organic Reference Standard, Carnosol
Catalog # 49702-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:5957-80-2
- Description:Carnosol
- Storage conditions:–20 °C
- Hazard class description:H317
- Shipping conditions:WICE
- MDL No.:MFCD02752467
- M-Clarity quality level:MQ100
- Synonym(s):Picrosalvin
- Molecular weight:330,42
Specifications
About this item
Caution: P280 - P302 + P352