Organic reference standard, Canthaxanthin (trans), analytical standard, Supelco®
Catalog # 32993-2MG
Supplier: Merck
CAS Number:
Specifications
- Pk:2 mg
- Standards:Analytical standard
- CAS No.:514-78-3
- Description:Canthaxanthin (trans)
- Melting point:201,6...201,7 °C
- Formula:C₄₀H₅₂O₂
- Storage temperature:2...8 °C
- Description:Canthaxanthin (trans)
- Form:Neat