Organic reference standard, Bisphenol A bis(3-chloro-2-hydroxypropyl) ether, analytical standard, Supelco®
Catalog # 15136-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:Analytical standard
- CAS No.:4809-35-2
- Description:Bisphenol A bis(3-chloro-2-hydroxypropyl) ether, ≥97,0% (HPLC)
- M-Clarity quality level:MQ100
- Formula:C₂₁H₂₆Cl₂O₄
- Description:Bisphenol A bis(3-chloro-2-hydroxypropyl) ether
- Form:Neat
Specifications
About this item
Standard for determining toxic monomers released from polymers of the inner coating of cans.