Organic reference standard, Bisphenol A (3-chloro-2-hydroxypropyl) (2,3-dihydroxypropyl) ether, analytical standard, Supelco®
Supplier: Merck
Certificates
About this item
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Specifications
- CAS No.:227947-06-0
- M-Clarity quality level:MQ100
- Formula:C₂₁H₂₇ClO₅
- Storage temperature:2...8 °C
- Description:Bisphenol A (3-chloro-2-hydroxypropyl) (2,3-dihydroxypropyl) ether
Specifications
Frequently Bought Together