Supelco® Organic Reference Standard, β-Sitosterol β-ᴅ-glucoside
Catalog # 89851-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:474-58-8
- Description:β-Sitosterol β-ᴅ-glucoside
- Storage conditions:2...30 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):β-Daucosterol; Coriandrinol; Daucosterin; Eleutheroside A
- Molecular weight:576,85
- Flash point:361 °C