Supelco® Organic Reference Standard, Bacopaside Ii
44698-5MG
: Merck
:
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:382146-66-9
- Description:Bacopaside II
- Storage conditions:2...30 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):Pseudojujubogenin 3-O-[α-ʟ-arabinofuranosyl-(1→2)-[β-ᴅ-glucopyranosyl-(1→3)]-β-ᴅ-glucopyranoside]
- Formula:C₄₇H₇₆O₁₈
- Molecular weight:929,10
- Flash point:Not applicable
- Form:Neat
: Bottomless glass bottle contents are inside inserted with fused cone.