Supelco® Organic Reference Standard, Lactose Monohydrate
47287-U
: Merck
:
- Pk:500 mg
- Standards:Analytical standard
- CAS No.:5989-81-1
- Description:Lactose monohydrate
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00150747
- M-Clarity quality level:MQ100
- Synonym(s):α-Lactose monohydrate; β-ᴅ-Gal-(1→4)-α-ᴅ-Glc; 4-O-β-ᴅ-Galactopyranosyl-α-ᴅ-glucose; Milk sugar
- Molecular weight:360,31