Organic reference standard, alpha-Carotene, analytical standard, Supelco®
Catalog # 50887-1MG
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mg
- Standards:Analytical standard
- CAS No.:7488-99-5
- Description:alpha-Carotene, ≥98,0% (HPLC)
- Melting point:183...185 °C
- Formula:C₄₀H₅₆
- Storage temperature:–20 °C
- Description:alpha-Carotene
- Form:Neat