Organic reference standard, Allura Red AC, analytical standard, Supelco®
Catalog # 38213-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:25956-17-6
- Description:Allura Red AC, ≥98,0% (HPLC)
- Melting point:>300 °C (lit.)
- Formula:C₁₈H₁₄N₂Na₂O₈S₂
- Description:Allura Red AC
- Form:Neat
Specifications
About this item
Allura Red AC comprises of 6-hydroxy-5-(2-methoxy-5-methyl-4-sulfonate-phenylazo)-2-napthalene-sulfonate, which is basically an azo-coupling product between the meta-isomer of cresidine-p-sulphonic acid (CSA) and Schaeffer′s salt (SS).
- It belongs to the class of monoazo dyes
- Soluble in water and is insoluble in ethanol
This powder exists in the form of dark red powder or granules and finds application in being used in food colors.
Packaging: Bottomless glass bottle contents are inside inserted with fused cone.