Organic reference standard, Aflatoxin G1 solution, analytical standard, Supelco®
Catalog # 34032-2ML-R
Supplier: Merck
CAS Number:
Specifications
- Concentration:2 µg/ml in acetonitrile
- Pk:2 mL
- Standards:Analytical standard
- CAS No.:1165-39-5
- Description:Aflatoxin G1 solution
- M-Clarity quality level:MQ100
- Formula:C₁₇H₁₂O₇
- Storage temperature:–20 °C
- Description:Aflatoxin G1 solution