Organic reference standard, 9-cis-Astaxanthin, analytical standard, Supelco®
Catalog # 51881-1MG
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mg
- Standards:Analytical standard
- CAS No.:113085-04-4
- Description:9-cis-Astaxanthin, ≤10% (all-trans-Astaxanthin, HPLC); ≥90,0% (HPLC)
- Formula:C₄₀H₅₂O₄
- Storage temperature:−20 °C
- Description:9-cis-Astaxanthin