Organic reference standard, 6-Ring NOGE (Novolac glycidyl ether) mixture of isomers, chain-like or branched, analytical standard, Supelco®
Catalog # 30977-50MG-F
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- Description:6-Ring NOGE (Novolac glycidyl ether) mixture of isomers, ≥80,0% (HPLC)
- M-Clarity quality level:MQ100
- Storage temperature:2...8 °C
- Description:6-Ring NOGE (Novolac glycidyl ether) mixture of isomers
- Form:Neat