Phytochemical standard, 6-Prenylnaringenin, analytical standard, Supelco®
75602-5MG
: Merck
:
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:68236-13-5
- Description:6-Prenylnaringenin, ≥95,0% (HPLC)
- Formula:C₂₀H₂₀O₅
- Storage temperature:2...8 °C
- Description:6-Prenylnaringenin
- Form:Neat
: Bottomless glass bottle. Contents are inside inserted fused cone.