Order Entry
Slovakia
ContactUsLinkComponent
Organic reference standard, 5-Pentadecylresorcinol, analytical standard, Supelco®
Organic reference standard, 5-Pentadecylresorcinol, analytical standard, Supelco®
Catalog # 91822-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 5-Pentadecylresorcinol, analytical standard, Supelco®
Catalog # 91822-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    3158-56-3
  • Description:
    5-Pentadecylresorcinol, ≥95,0% (HPLC)
  • Formula:
    C₂₁H₃₆O₂
  • Storage temperature:
    2...8 °C
  • Description:
    5-Pentadecylresorcinol
  • Form:
    Neat

Specifications

About this item

5-Pentadecylresorcinol is a resorcinol derivative, that can be found in the wood of Grevillea banksii.

Occurs in peel and flesh of unripe mangos, Mangifera indica (Anacardiaceae).

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.