Organic reference standard, 5-Heptadecylresorcinol, analytical standard, Supelco®
97001-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:70110-59-7
- Description:5-Heptadecylresorcinol, ≥95,0% (HPLC)
- Formula:C₂₃H₄₀O₂
- Storage temperature:2…8 °C
- Description:5-Heptadecylresorcinol
- Form:Neat
: Bottomless glass bottle. Contents are inside inserted fused cone.