Organic reference standard, 3-Ring NOGE (Novolac glycidyl ether) mixture of isomers, analytical standard, Supelco®
68931-50MG-F
: Merck
:
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:158163-01-0
- Description:3-Ring NOGE (Novolac glycidyl ether) mixture of isomers, ≥90,0% (HPLC)
- M-Clarity quality level:MQ100
- Storage temperature:2...8 °C
- Description:3-Ring NOGE (Novolac glycidyl ether) mixture of isomers
- Form:Neat