Organic reference standard, 3-Propylidenephthalide, mixture of isomers, analytical standard, Supelco®
75012-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:17369-59-4
- Description:3-Propylidenephthalide, mixture of isomers, ≥98,0% (sum of isomers, GC)
- Formula:C₁₁H₁₀O
- Description:3-Propylidenephthalide