Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, 3-O-Acetyl-11-keto-β-boswellic acid
  74607-5MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, 3-O-Acetyl-11-keto-β-boswellic acid
  74607-5MG
 :  Merck
 :  

 

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    67416-61-9
  • Description:
    3-O-Acetyl-11-keto-β-boswellic acid
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD03788777
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    11-keto-β-Boswellic acid acetate; 3α-Acetoxy-11-oxo-12-ursen-24-oic acid; AKBA
  • Molecular weight:
    512,72