Organic reference standard, 3-Acetyldeoxynivalenol, Supelco®
32927-5MG
: Merck
:
- Pk:5 mg
- CAS No.:50722-38-8
- Description:3-Acetyldeoxynivalenol
- M-Clarity quality level:MQ100
- Formula:C₁₇H₂₂O₇
- Description:3-Acetyldeoxynivalenol
- Flash point:44 °C
- Boiling point:130 °C mmHg (lit.)
- Form:Neat
3-Acetyldeoxynivalenol belongs to the class of trichothecenes, which are secondary metabolites generated by various fungi, particularly by Fusarium species.
3-Acetyldeoxynivalenol may be used as an analytical reference standard for the determination of the analyte in: Cereals by gas chromatography-mass spectrometry (GC-MS), milk samples by liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).