Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, 3,3′-Diindolylmethane
  74601-100MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, 3,3′-Diindolylmethane
  74601-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    1968-05-4
  • Description:
    3,3′-Diindolylmethane
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H315 - H319 - H335 - H413
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00195766
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3,3′-Bisindolylmethane; DIM
  • Molecular weight:
    246,31

 

 

  • GHS signal: GHS07
 : P273 - P302 + P352 - P305 + P351 + P338