Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, 1-O-Galloyl-β-ᴅ-glucose
Supelco® Organic Reference Standard, 1-O-Galloyl-β-ᴅ-glucose
Catalog # 69288-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 1-O-Galloyl-β-ᴅ-glucose
Catalog # 69288-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    13405-60-2
  • Description:
    1-O-Galloyl-β-ᴅ-glucose
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD11975493
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    β-ᴅ-Glucopyranose 1-(3,4,5-trihydroxybenzoate); β-Glucogallin; 1-O-Galloyl-β-ᴅ-glucopyranose
  • Molecular weight:
    332,26

Specifications

About this item

Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338