Supelco® Organic Reference Standard, 1,3,6-Tri-O-galloyl-β-ᴅ-glucose
Catalog # 78864-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:18483-17-5
- Description:1,3,6-Tri-O-galloyl-β-ᴅ-glucose
- Storage conditions:2...30 °C
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD06656305
- M-Clarity quality level:MQ100
- Synonym(s):β-ᴅ-Glucopyranose 1,3,6-trigallate; β-ᴅ-Glucopyranose-1,3,6-tris(3,4,5-trihydroxybenzoate); 1,3,6-Tri-O-galloyl-β-ᴅ-glucopyranose; NP 005114; NSC 69861
- Molecular weight:636,47
Specifications
About this item
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338