Supelco® Organic Reference Standard, [6]-Shogaol
39303-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:555-66-8
- Description:[6]-Shogaol
- Storage conditions:2...8 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD01736094
- M-Clarity quality level:MQ100
- Synonym(s):1-(4-Hydroxy-3-methoxyphenyl)-4-decen-3-one
- Formula:C₁₇H₂₄O₃
- Molecular weight:276,37
- Storage temperature:2…8 °C
- Flash point:Not applicable
- Form:Neat
- GHS signal: GHS07
: Bottomless glass bottle. Contents are inside inserted fused cone.