Supelco® Organic Reference Standard, (1r)-(−)-Menthyl Acetate
Supplier: Merck
Certificates
About this item
Specifications
- CAS No.:2623-23-6
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00001482
- M-Clarity quality level:MQ100
- Formula:C₁₂H₂₂O₂
- Molecular weight:198,30
- Density:0,92 g/ml at 25 °C (lit.)
- Flash point:77 °C - closed cup
- Boiling point:229...230 °C (lit.)
Specifications
Frequently Bought Together