Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, (+)-Terpinen-4-ol
Supelco® Organic Reference Standard, (+)-Terpinen-4-ol
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, (+)-Terpinen-4-ol
Supplier:  Merck
Certificates

Specifications

  • CAS No.:
    2438-10-0
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302 - H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00077431
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (S)-1-Isopropyl-4-methyl-3-cyclohexen-1-ol; (S)-p-Menth-1-en-4-ol
  • Molecular weight:
    154,25
  • Density:
    0,933 g/ml at 20 °C (lit.)
  • Flash point:
    79 °C - Closed cup
  • Boiling point:
    211 - 213 °C (lit.)

Specifications

Related Information

Frequently Bought Together