Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, (+)-Catechin
  43412-10MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, (+)-Catechin
  43412-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    154-23-4
  • Description:
    (+)-Catechin
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H315 - H319 - H335
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00075649
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (+)-Cyanidanol; ᴅ-Catechin
  • Formula:
    C₁₅H₁₄O₆
  • Molecular weight:
    290,27
  • Storage temperature:
    2…8 °C
  • Flash point:
    Not applicable
  • Form:
    Neat

 

 

 : Bottomless glass bottle. Contents are inside inserted fused cone.
 : P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338