Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, (−)-Gallocatechin gallate
  80352-10MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, (−)-Gallocatechin gallate
  80352-10MG
 :  Merck
 :  

 

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    4233-96-9
  • Description:
    (−)-Gallocatechin gallate
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H315 - H319 - H335
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00214298
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (2S,3R)-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol 3-(3,4,5-trihydroxybenzoate)
  • Molecular weight:
    458,37

 

 

  • GHS signal: GHS07
 : P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338