Supelco® Organic Reference Standard, (−)-Calcium hydroxycitrate tribasic
Catalog # 75450-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:921226-01-9
- Description:(−)-Calcium hydroxycitrate tribasic
- Storage conditions:2...30 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):(−)-Hydroxycitric acid tricalcium salt
- Molecular weight:530,43