Phytochemical standard, (-)-alpha-Santonin, analytical standard, Supelco®
Catalog # 53653-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:481-06-1
- Description:(-)-alpha-Santonin, ≥95,0% (HPLC)
- Melting point:172...173 °C (lit.)
- Formula:C₁₅H₁₈O₃
- Storage temperature:2…8 °C
- Description:(-)-alpha-Santonin
- Form:Neat
Specifications
About this item
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.