Supelco® Organic Reference Standard, trans-11-Vaccenic methyl ester
CRM46905
: Merck
:
- Concentration:10 mg/ml in heptane
- Pk:1 mL
- Standards:Certified reference material
- Description:trans-11-Vaccenic methyl ester
- Storage conditions:–10...–25 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:DICE
- M-Clarity quality level:MQ100
- Flash point:–3,99 °C – Closed cup
- Pk Info:Ampule of 1 ml
- GHS signal: GHS02, GHS08, GHS07, GHS09
: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331