Supelco® Organic Reference Standard, Hydrocarbon Mix
49137-U
: Merck
:
- Concentration:1000 μg/ml each component in cyclohexane
- Pk:1
- Standards:Certified reference material
- Description:Hydrocarbon mix
- Storage conditions:2…8 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:–18 °C
- GHS signal: GHS02, GHS08, GHS07, GHS09
: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331