Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, ASTM® D5580 Quantitative Calibration Kit
Supelco® Organic Reference Standard, ASTM® D5580 Quantitative Calibration Kit
Catalog # 47734-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, ASTM® D5580 Quantitative Calibration Kit
Catalog # 47734-U
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Standards:
    Analytical standard
  • Description:
    ASTM® D5580 quantitative calibration kit
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H225 - H301 + H311 + H331 - H304 - H315 - H319 - H336 - H340 - H350 - H361fd - H370 - H372 - H410 - H420
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Flash point:
    –12 °C

Specifications

About this item

  • GHS signal: GHS02, GHS06, GHS08, GHS09
Caution: P210 - P273 - P280 - P301 + P310 - P331 - P502