Supelco® Organic Reference Standard, Fatty Acids Unsaturated Kit
Catalog # UN10-1KT
Supplier: Merck
CAS Number:
Specifications
- Pk:1 KIT
- Standards:Analytical standard
- Description:Fatty acids unsaturated kit
- Storage conditions:–20 °C
- Hazard class description:H315 - H319 - H335 - H413
- Shipping conditions:DICE
- M-Clarity quality level:MQ100
- Flash point:62 °C
Specifications
About this item
- GHS signal: GHS07
Delivery: Analytes: Arachidonic acid 100 mg; cis-4,7,10,13,16,19-Docosahexaenoic acid 100 mg; Elaidic acid 100 mg; Erucic acid 100 mg; Linoleic acid 100 mg; Linolenic acid 100 mg; Nervonic acid 100 mg; Oleic acid 100 mg; Palmitoleic acid 100 mg; Petroselinic acid 100 mg.
Caution: P261 - P264 - P271 - P273 - P302 + P352 - P305 + P351 + P338