Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Fatty Acid Methyl Esters, Unsaturated Kit
Supelco® Organic Reference Standard, Fatty Acid Methyl Esters, Unsaturated Kit
Catalog # ME14-1KT
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Fatty Acid Methyl Esters, Unsaturated Kit
Catalog # ME14-1KT
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Standards:
    Analytical standard
  • Description:
    Fatty acid methyl esters, unsaturated kit
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H315 - H319 - H335
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100

Specifications

About this item

  • GHS signal: GHS07
Delivery: Analytes: cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester 100 mg; Methyl arachidonate 100 mg; Methyl cis-13-docosenoate 100 mg; Methyl cis-11-eicosenoate 100 mg; Methyl elaidate 100 mg; Methyl linoleate 100 mg; Methyl linolelaidate 100 mg; Methyl linolenate 100 mg; Methyl myristoleate 100 mg; Methyl oleate 100 mg; Methyl palmitoleate 100 mg; Methyl petroselinate 100 mg; Methyl cis-15-tetracosenoate 100 mg.
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338