Supelco® Organic Reference Standard, Fatty Acid Kit
EC10A-1KT
: Merck
:
- Pk:1 KIT
- Standards:Analytical standard
- Description:Fatty Acid Kit
- Storage conditions:2…8 °C
- Hazard class description:H302 - H311 - H314 - H412
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- GHS signal: GHS05
: P273 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P310 - P305 + P351 + P338