Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Escherichia coli ATCC 11303 cells
Supelco® PESTANAL® Organic Reference Standard, Escherichia coli ATCC 11303 cells
  45704-100MG-R
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Escherichia coli ATCC 11303 cells
  45704-100MG-R
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, analytical standard
  • Description:
    Escherichia coli ATCC 11303 cells
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00211924
  • M-Clarity quality level:
    MQ100

 

 

  • GHS signal: GHS07, GHS09