Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Trietazine
  31761-100MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Trietazine
  31761-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    1912-26-1
  • Description:
    Trietazine
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H400
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00128251
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₉H₁₆ClN₅
  • Molecular weight:
    229,71
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS07, GHS09
 : P264 - P270 - P273 - P301 + P312 - P391 - P501