Supelco® PESTANAL® Organic Reference Standard, Transfluthrin
Catalog # 46114-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:118712-89-3
- Description:Transfluthrin
- Storage conditions:2…8 °C
- Hazard class description:H315 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Molecular weight:371,15
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P264 - P273 - P280 - P302 + P352 - P332 + P313 - P362 + P364