Supelco® PESTANAL® Organic Reference Standard, TCPP, mixture of isomers
Catalog # 32952-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:13674-84-5
- Description:TCPP, mixture of isomers
- Storage conditions:2...30 °C
- Hazard class description:H302
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Formula:C₉H₁₈Cl₃O₄P
- Molecular weight:327,57
- Flash point:185 °C - closed cup
Specifications
About this item
- GHS signal: GHS07
Caution: P264 - P270 - P301 + P312 - P501