Organic reference standard, Sulisobenzone, analytical standard, Supelco®
50194-100MG
: Merck
:
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:4065-45-6
- Description:Sulisobenzone
- Formula:C₁₄H₁₂O₆S
- Description:Sulisobenzone
: Bottomless glass bottle. Contents are inside inserted fused cone.