Supelco® PESTANAL® Organic Reference Standard, Sulcotrione
46318-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:99105-77-8
- Description:Sulcotrione
- Storage conditions:2...30 °C
- Hazard class description:H317 - H361d - H373 - H410
- Shipping conditions:STD
- MDL No.:MFCD01632349
- M-Clarity quality level:MQ100
- Synonym(s):2-[2-Chloro-4-(methylsulfonyl)benzoyl]-1,3-cyclohexanedione
- Molecular weight:328,77
- GHS signal: GHS08, GHS09
: P202 - P260 - P273 - P280 - P302 + P352 - P308 + P313