Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Secbumeton
Supelco® PESTANAL® Organic Reference Standard, Secbumeton
  45658-250MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Secbumeton
  45658-250MG
 :  Merck
 :  

 

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    26259-45-0
  • Description:
    Secbumeton
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 + H312 - H319 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00078683
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₀H₁₉N₅O
  • Molecular weight:
    225,29
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS07, GHS09
 : P273 - P280 - P301 + P312 + P330 - P302 + P352 + P312 - P305 + P351 + P338