Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Rimsulfuron
Supelco® PESTANAL® Organic Reference Standard, Rimsulfuron
Catalog # 31658-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Rimsulfuron
Catalog # 31658-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    122931-48-0
  • Description:
    Rimsulfuron
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD01632305
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1-(4,6-Dimethoxy-2-pyrimidinyl)-3-[3-(ethylsulfonyl)-2-pyridylsulfonyl]urea
  • Formula:
    C₁₄H₁₇N₅O₇S₂
  • Molecular weight:
    431,44
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07